Assay Details
| Gene Symbol: Fbrsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142642.1 | NP_001136114.1 | 10 - 11 | 1873 | 63 | |
| NM_028596.2 | NP_082872.2 | 4 - 5 | 989 | 63 | ||
| GenBank mRNA | AK031967.1 | - | 4 - 5 | 959 | 63 | |
| BC151196.1 | - | 5 - 6 | 882 | 9088084 | 63 | |
| BU614458.1 | - | 1 - 2 | 181 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fbrsl1
Genomic Map