Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500009850.mm.1 |
TC0500001146.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500009850.mm.1 |
NM_001122639.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009850.mm.1 |
NM_198306.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009850.mm.1 |
NR_104449.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009849.mm.1 |
TC0500001146.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500009849.mm.1 |
NM_001122639.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009849.mm.1 |
NM_198306.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500009849.mm.1 |
NR_104449.1 |
Refseq |