Assay Details
| Gene Symbol: Tctn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039153.3 | NP_001034242.2 | 11 - 12 | 1453 | 119 | |
| NR_104441.1 | 10 - 11 | 1376 | 119 | |||
| GenBank mRNA | AK036568.1 | - | 11 - 12 | 1435 | 119 | |
| AK138661.1 | - | 10 - 11 | 1375 | 119 | ||
| AK144772.1 | - | 11 - 12 | 1429 | 119 | ||
| DQ278867.1 | - | 11 - 12 | 1454 | 119 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tctn1
Genomic Map