Assay Details
| Gene Symbol: Wscd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177292.3 | NP_796266.2 | 1827 | 117 | ||
| GenBank mRNA | BC096526.1 | - | 1827 | 117 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Wscd2
Genomic Map