Assay Details
| Gene Symbol: Tnrc18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122730.2 | NP_001116202.2 | 16 - 17 | 5746 | 76 | |
| GenBank mRNA | AK141849.1 | - | 13 - 14 | 4845 | 76 | |
| CB249572.1 | - | 1 - 2 | 167 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tnrc18
Genomic Map