Assay Details
| Gene Symbol: Tnrc18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122730.2 | NP_001116202.2 | 17 - 18 | 6174 | 102 | |
| GenBank mRNA | CB249572.1 | - | 2 - 3 | 595 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tnrc18
Genomic Map