Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500032403.mm.1 |
TC0500003533.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500032403.mm.1 |
NM_001122730.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500032403.mm.1 |
NM_178242.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500032403.mm.1 |
NM_178242.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500032404.mm.1 |
TC0500003533.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500032404.mm.1 |
NM_001122730.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500032404.mm.1 |
NM_178242.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500032404.mm.1 |
NM_178242.2 |
Refseq |