Assay Details
| Gene Symbol: Tnrc18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122730.2 | NP_001116202.2 | 6 - 7 | 2609 | 74 | |
| NM_178242.2 | NP_839973.1 | 6 - 7 | 2609 | 74 | ||
| GenBank mRNA | AK141849.1 | - | 3 - 4 | 1701 | 74 | |
| AK173280.1 | - | 3 - 4 | 364 | 74 | ||
| BC025631.1 | - | 1 - 2 | 139 | 74 | ||
| BC049818.1 | - | 6 - 7 | 2609 | 6511384 | 74 | |
| BC115863.1 | - | 2 - 3 | 255 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tnrc18
Genomic Map