Assay Details
| Gene Symbol: Tnrc18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122730.2 | NP_001116202.2 | 7 - 8 | 2752 | 99 | |
| NM_178242.2 | NP_839973.1 | 7 - 8 | 2752 | 99 | ||
| GenBank mRNA | AK173280.1 | - | 4 - 5 | 507 | 99 | |
| BC025631.1 | - | 2 - 3 | 282 | 99 | ||
| BC049818.1 | - | 7 - 8 | 2752 | 6511384 | 99 | |
| BC115863.1 | - | 3 - 4 | 398 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tnrc18
Genomic Map