Assay Details
| Gene Symbol: Mrps17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025450.4 | NP_079726.1 | 2 - 3 | 280 | 89 | |
| GenBank mRNA | AK007356.1 | - | 2 - 3 | 281 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK007356.1, NM_025450.4 transcripts.
Set Membership: |
|

Mrps17
Genomic Map