Assay Details
| Gene Symbol: Tfr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289507.1 | NP_001276436.1 | 15 - 16 | 1955 | 82 | |
| NM_001289509.1 | NP_001276438.1 | 17 - 18 | 1974 | 82 | ||
| NM_001289511.1 | NP_001276440.1 | 16 - 17 | 1832 | 82 | ||
| NM_015799.4 | NP_056614.3 | 17 - 18 | 1914 | 82 | ||
| GenBank mRNA | AF207741.1 | - | 15 - 16 | 1789 | 82 | |
| AK004848.2 | - | 17 - 18 | 1890 | 82 | ||
| AK004965.1 | - | 17 - 18 | 1945 | 82 | ||
| AK155526.1 | - | 16 - 17 | 2052 | 82 | ||
| AK165320.1 | - | 15 - 16 | 1783 | 82 | ||
| BC013654.1 | - | 15 - 16 | 1955 | 4196597 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tfr2
Genomic Map