Assay Details
| Gene Symbol: Mvk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF137598.1 | - | 3 - 4 | 392 | 81 | |
| AK009261.1 | - | 5 - 6 | 502 | 81 | ||
| AK031084.1 | - | 3 - 4 | 1173 | 81 | ||
| AK149630.1 | - | 4 - 5 | 489 | 81 | ||
| BC005606.1 | - | 4 - 5 | 445 | 3498140 | 81 | |
More Information
Set Membership: |
|

Mvk
Genomic Map