Assay Details
| Gene Symbol: Gstm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010358.5 | NP_034488.1 | 1 - 2 | 235 | 80 | |
| GenBank mRNA | BC003822.1 | - | 1 - 2 | 58 | 3600444 | 80 |
| BC046758.1 | - | 1 - 2 | 88 | 5693552 | 80 | |
| BC091763.1 | - | 1 - 2 | 68 | 5102093 | 80 | |
| J03952.1 | - | 1 - 2 | 49 | 80 | ||
| J04632.1 | - | 1 - 2 | 236 | 80 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC003822.1, BC046758.1, BC091763.1, J03952.1, J04632.1, NM_010358.5 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gstm1
Genomic Map