Assay Details
| Gene Symbol: Dtx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008052.3 | NP_032078.2 | 6 - 7 | 2090 | 65 | |
| GenBank mRNA | AB015422.1 | - | 5 - 6 | 1693 | 65 | |
| AK088630.1 | - | 6 - 7 | 2046 | 65 | ||
| AK169455.1 | - | 6 - 7 | 2091 | 65 | ||
| AK220514.1 | - | 5 - 6 | 1671 | 65 | ||
| BC053055.1 | - | 6 - 7 | 1236 | 5685525 | 65 | |
| BC065075.1 | - | 7 - 8 | 1872 | 65 | ||
| U38252.1 | - | 4 - 5 | 1275 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dtx1
Genomic Map