Assay Details
| Gene Symbol: Dtx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008052.3 | NP_032078.2 | 8 - 9 | 2419 | 100 | |
| GenBank mRNA | AB015422.1 | - | 7 - 8 | 2022 | 100 | |
| AK088630.1 | - | 8 - 9 | 2375 | 100 | ||
| AK169455.1 | - | 8 - 9 | 2420 | 100 | ||
| AK220514.1 | - | 7 - 8 | 2000 | 100 | ||
| BC053055.1 | - | 8 - 9 | 1565 | 5685525 | 100 | |
| BC065075.1 | - | 9 - 10 | 2201 | 100 | ||
| U38252.1 | - | 6 - 7 | 1604 | 100 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dtx1
Genomic Map