Assay Details
| Gene Symbol: Scd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024450.2 | NP_077770.1 | 1 - 2 | 263 | 66 | |
| GenBank mRNA | AF272037.1 | - | 1 - 2 | 252 | 66 | |
| AK078675.1 | - | 1 - 2 | 284 | 66 | ||
| AK078713.1 | - | 1 - 2 | 285 | 66 | ||
| AK136277.1 | - | 1 - 2 | 280 | 66 | ||
| BC118033.1 | - | 1 - 2 | 161 | 40106893 | 66 | |
| BC118045.1 | - | 1 - 2 | 161 | 40106896 | 66 | |
More Information
Set Membership: |
|

Scd3
Genomic Map