Assay Details
| Gene Symbol: Sufu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025391.2 | NP_001020562.1 | 8 - 9 | 1231 | 77 | |
| NM_015752.3 | NP_056567.2 | 8 - 9 | 1234 | 77 | ||
| GenBank mRNA | AF134893.1 | - | 8 - 9 | 1174 | 77 | |
| AJ131692.1 | - | 8 - 9 | 1232 | 77 | ||
| AJ308625.1 | - | 8 - 9 | 910 | 77 | ||
| AJ308626.1 | - | 8 - 9 | 1165 | 77 | ||
| AK047603.1 | - | 8 - 9 | 1104 | 77 | ||
| AK156410.1 | - | 8 - 9 | 1196 | 77 | ||
| BC048168.1 | - | 8 - 9 | 1204 | 6821477 | 77 | |
| BC056997.1 | - | 8 - 9 | 1204 | 6821477 | 77 | |
More Information
Set Membership: |
|

Sufu
Genomic Map