Assay Details
| Gene Symbol: Sufu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025391.2 | NP_001020562.1 | 9 - 10 | 1363 | 75 | |
| NM_015752.3 | NP_056567.2 | 9 - 10 | 1366 | 75 | ||
| GenBank mRNA | AF134893.1 | - | 9 - 10 | 1306 | 75 | |
| AJ131692.1 | - | 9 - 10 | 1364 | 75 | ||
| AJ308626.1 | - | 9 - 10 | 1297 | 75 | ||
| AK016976.1 | - | 9 - 10 | 1344 | 75 | ||
| AK047603.1 | - | 9 - 10 | 1236 | 75 | ||
| AK156410.1 | - | 9 - 10 | 1328 | 75 | ||
| BC048168.1 | - | 9 - 10 | 1336 | 6821477 | 75 | |
| BC056997.1 | - | 9 - 10 | 1336 | 6821477 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sufu
Genomic Map