Assay Details
| Gene Symbol: Ddx55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190795.1 | NP_001177724.1 | 10 - 11 | 1118 | 110 | |
| NM_026409.4 | NP_080685.2 | 10 - 11 | 1118 | 110 | ||
| GenBank mRNA | AK012782.1 | - | 10 - 11 | 1098 | 110 | |
| AK017554.1 | - | 10 - 11 | 1060 | 110 | ||
| AK033741.1 | - | 10 - 11 | 1095 | 110 | ||
| AK077525.1 | - | 10 - 11 | 1099 | 110 | ||
| AK129402.1 | - | 10 - 11 | 1096 | 110 | ||
| AK154422.1 | - | 10 - 11 | 1119 | 110 | ||
| AK167149.1 | - | 9 - 10 | 3113 | 110 | ||
| BC043052.1 | - | 10 - 11 | 1068 | 110 | ||
| BC117786.1 | - | 10 - 11 | 1080 | 40092124 | 110 | |
| BC117787.1 | - | 10 - 11 | 1080 | 40092125 | 110 | |
More Information
Set Membership: |
|

Ddx55
Genomic Map