Assay Details
| Gene Symbol: Ddx55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026409.4 | NP_080685.2 | 11 - 12 | 1234 | 74 | |
| GenBank mRNA | AK012782.1 | - | 11 - 12 | 1214 | 74 | |
| AK017554.1 | - | 11 - 12 | 1176 | 74 | ||
| AK033741.1 | - | 11 - 12 | 1211 | 74 | ||
| AK077525.1 | - | 11 - 12 | 1215 | 74 | ||
| AK129402.1 | - | 11 - 12 | 1212 | 74 | ||
| AK154422.1 | - | 11 - 12 | 1235 | 74 | ||
| AK167149.1 | - | 10 - 11 | 3229 | 74 | ||
| BC025218.1 | - | 1 - 2 | 67 | 74 | ||
| BC117786.1 | - | 11 - 12 | 1196 | 40092124 | 74 | |
| BC117787.1 | - | 11 - 12 | 1196 | 40092125 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ddx55
Genomic Map