Assay Details
| Gene Symbol: Ddx55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190795.1 | NP_001177724.1 | 7 - 8 | 809 | 63 | |
| NM_026409.4 | NP_080685.2 | 7 - 8 | 809 | 63 | ||
| GenBank mRNA | AK012782.1 | - | 7 - 8 | 789 | 63 | |
| AK033741.1 | - | 7 - 8 | 786 | 63 | ||
| AK077525.1 | - | 7 - 8 | 790 | 63 | ||
| AK129402.1 | - | 7 - 8 | 787 | 63 | ||
| AK154422.1 | - | 7 - 8 | 810 | 63 | ||
| AK167149.1 | - | 6 - 7 | 2804 | 63 | ||
| BC043052.1 | - | 7 - 8 | 759 | 63 | ||
| BC117786.1 | - | 7 - 8 | 771 | 40092124 | 63 | |
| BC117787.1 | - | 7 - 8 | 771 | 40092125 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ddx55
Genomic Map