Assay Details
| Gene Symbol: Abcg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030239.2 | NP_084515.2 | 10 - 11 | 1445 | 82 | |
| GenBank mRNA | AF324242.1 | - | 10 - 11 | 1397 | 82 | |
| AK030831.1 | - | 10 - 11 | 1447 | 82 | ||
| AK051880.1 | - | 10 - 11 | 1411 | 82 | ||
| AK156626.1 | - | 10 - 11 | 1413 | 82 | ||
| BC138772.1 | - | 10 - 11 | 1426 | 8861794 | 82 | |
| BC138773.1 | - | 10 - 11 | 1425 | 8861795 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Abcg3
Genomic Map