Assay Details
| Gene Symbol: Abcg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030239.2 | NP_084515.2 | 13 - 14 | 1806 | 78 | |
| GenBank mRNA | AK030831.1 | - | 13 - 14 | 1808 | 78 | |
| AK051880.1 | - | 13 - 14 | 1772 | 78 | ||
| AK156626.1 | - | 13 - 14 | 1774 | 79 | ||
| BC029095.1 | - | 11 - 12 | 1561 | 78 | ||
| BC138772.1 | - | 13 - 14 | 1787 | 8861794 | 78 | |
| BC138773.1 | - | 13 - 14 | 1786 | 8861795 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Abcg3
Genomic Map