Assay Details
| Gene Symbol: Pkd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008861.3 | NP_032887.3 | 10 - 11 | 2294 | 106 | |
| GenBank mRNA | AF014010.1 | - | 10 - 11 | 2197 | 106 | |
| AK053502.1 | - | 10 - 11 | 2199 | 106 | ||
| AK128961.1 | - | 11 - 12 | 2068 | 106 | ||
| AK144421.1 | - | 1 - 2 | 2222 | 106 | ||
| BC053058.1 | - | 10 - 11 | 2301 | 106 | ||
| BC062969.1 | - | 10 - 11 | 2301 | 106 | ||
| FJ609779.1 | - | 9 - 10 | 2042 | 106 | ||
| Y13278.1 | - | 10 - 11 | 2210 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pkd2
Genomic Map