Assay Details
| Gene Symbol: Pkd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008861.3 | NP_032887.3 | 11 - 12 | 2416 | 61 | |
| GenBank mRNA | AF014010.1 | - | 11 - 12 | 2319 | 61 | |
| AK053502.1 | - | 11 - 12 | 2321 | 61 | ||
| AK128961.1 | - | 12 - 13 | 2190 | 61 | ||
| BC053058.1 | - | 11 - 12 | 2423 | 61 | ||
| BC062969.1 | - | 11 - 12 | 2423 | 61 | ||
| CX566027.1 | - | 2 - 3 | 143 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pkd2
Genomic Map