Assay Details
| Gene Symbol: Pkd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008861.3 | NP_032887.3 | 13 - 14 | 2693 | 162 | |
| GenBank mRNA | AF014010.1 | - | 13 - 14 | 2596 | 162 | |
| AK053502.1 | - | 13 - 14 | 2598 | 162 | ||
| AK128961.1 | - | 14 - 15 | 2467 | 162 | ||
| BC053058.1 | - | 13 - 14 | 2700 | 162 | ||
| BC062969.1 | - | 13 - 14 | 2700 | 162 | ||
| CX566027.1 | - | 4 - 5 | 420 | 162 | ||
| FJ609779.1 | - | 12 - 13 | 2441 | 162 | ||
| Y13278.1 | - | 13 - 14 | 2609 | 162 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pkd2
Genomic Map