Assay Details
| Gene Symbol: Pkd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008861.3 | NP_032887.3 | 4 - 5 | 1268 | 70 | |
| GenBank mRNA | AK053502.1 | - | 4 - 5 | 1173 | 70 | |
| AK164673.1 | - | 4 - 5 | 1159 | 70 | ||
| BC053058.1 | - | 4 - 5 | 1275 | 70 | ||
| BC062969.1 | - | 4 - 5 | 1275 | 70 | ||
| FJ609779.1 | - | 4 - 5 | 1184 | 70 | ||
| Y13278.1 | - | 4 - 5 | 1184 | 70 | ||
| Y14120.1 | - | 4 - 5 | 934 | 70 | ||
More Information
Set Membership: |
|

Pkd2
Genomic Map