Assay Details
| Gene Symbol: Pkd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008861.3 | NP_032887.3 | 8 - 9 | 2075 | 76 | |
| GenBank mRNA | AF014010.1 | - | 8 - 9 | 1978 | 76 | |
| AK053502.1 | - | 8 - 9 | 1980 | 76 | ||
| AK128961.1 | - | 9 - 10 | 1849 | 76 | ||
| BC053058.1 | - | 8 - 9 | 2082 | 76 | ||
| BC062969.1 | - | 8 - 9 | 2082 | 76 | ||
| FJ609779.1 | - | 7 - 8 | 1823 | 76 | ||
| Y13278.1 | - | 8 - 9 | 1991 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pkd2
Genomic Map