Assay Details
| Gene Symbol: Dgat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010046.2 | NP_034176.1 | 15 - 16 | 1319 | 86 | |
| GenBank mRNA | AB057816.1 | - | 15 - 16 | 1307 | 86 | |
| AF078752.1 | - | 15 - 16 | 1377 | 86 | ||
| AK008995.1 | - | 2 - 3 | 447 | 86 | ||
| BC003717.1 | - | 15 - 16 | 1319 | 3486724 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dgat1
Genomic Map