Assay Details
| Gene Symbol: Dgat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010046.2 | NP_034176.1 | 4 - 5 | 475 | 85 | |
| GenBank mRNA | AB057816.1 | - | 4 - 5 | 463 | 85 | |
| BC003717.1 | - | 4 - 5 | 475 | 3486724 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dgat1
Genomic Map