Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500013909.mm.1 |
NM_001168288.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013909.mm.1 |
NM_001168288.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013909.mm.1 |
NM_198420.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013909.mm.1 |
NM_198420.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013909.mm.1 |
TC1500001791.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500013910.mm.1 |
NM_001168288.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013910.mm.1 |
NM_001168288.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013910.mm.1 |
NM_198420.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013910.mm.1 |
NM_198420.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013910.mm.1 |
TC1500001791.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500013911.mm.1 |
NM_001168288.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013911.mm.1 |
NM_001168288.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013911.mm.1 |
NM_198420.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013911.mm.1 |
NM_198420.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500013911.mm.1 |
TC1500001791.mm.1 |
MTA TC |