Assay Details
| Gene Symbol: Arhgap39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168288.1 | NP_001161760.1 | 7 - 8 | 2858 | 74 | |
| GenBank mRNA | AK035479.1 | - | 6 - 7 | 2563 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap39
Genomic Map