Assay Details
| Gene Symbol: Arhgap39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168288.1 | NP_001161760.1 | 8 - 9 | 2937 | 66 | |
| NM_198420.2 | NP_940812.2 | 7 - 8 | 2844 | 66 | ||
| GenBank mRNA | AK035479.1 | - | 7 - 8 | 2642 | 66 | |
| AK173232.1 | - | 7 - 8 | 2779 | 66 | ||
| BC060065.1 | - | 3 - 4 | 1416 | 66 | ||
| BC060637.1 | - | 7 - 8 | 2825 | 6849400 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgap39
Genomic Map