Assay Details
| Gene Symbol: Tonsl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183091.3 | NP_898914.3 | 21 - 22 | 3401 | 90 | |
| GenBank mRNA | BC066068.1 | - | 21 - 22 | 3383 | 6851399 | 90 |
More Information
Set Membership: |
|

Tonsl
Genomic Map