Assay Details
Gene Symbol: Emc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_197991.2 | NP_932108.2 | 3 - 4 | 439 | 73 | |
GenBank mRNA | AK009801.1 | - | 3 - 4 | 349 | 72 | |
AK150499.1 | - | 3 - 4 | 344 | 73 | ||
AK160274.1 | - | 3 - 4 | 354 | 73 | ||
AK169010.1 | - | 3 - 4 | 390 | 73 | ||
AK171657.1 | - | 3 - 4 | 353 | 73 | ||
AY761096.1 | - | 3 - 4 | 332 | 73 | ||
AY761098.1 | - | 3 - 4 | 332 | 73 | ||
BC006065.1 | - | 1 - 2 | 76 | 73 | ||
BC020179.1 | - | 3 - 4 | 334 | 4913500 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
