Assay Details
Gene Symbol: Emc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_197991.2 | NP_932108.2 | 4 - 5 | 545 | 65 | |
GenBank mRNA | AK150499.1 | - | 4 - 5 | 450 | 65 | |
AK169010.1 | - | 4 - 5 | 496 | 65 | ||
AK171657.1 | - | 4 - 5 | 459 | 65 | ||
AY761096.1 | - | 4 - 5 | 438 | 65 | ||
AY761098.1 | - | 4 - 5 | 438 | 65 | ||
BC006065.1 | - | 2 - 3 | 182 | 65 | ||
BC020179.1 | - | 4 - 5 | 440 | 4913500 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
