Assay Details
Gene Symbol: Emc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY761096.1 | - | 6 - 7 | 720 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
