Assay Details
| Gene Symbol: Patl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172635.3 | NP_766223.2 | 11 - 12 | 1594 | 63 | |
| GenBank mRNA | AK085013.1 | - | 11 - 12 | 1536 | 63 | |
| AK087559.1 | - | 11 - 12 | 1558 | 63 | ||
| AK150301.1 | - | 11 - 12 | 1595 | 63 | ||
| AK154873.1 | - | 11 - 12 | 1546 | 63 | ||
| AK161738.1 | - | 5 - 6 | 556 | 63 | ||
| AK170408.1 | - | 11 - 12 | 1558 | 63 | ||
| AK170745.1 | - | 11 - 12 | 1559 | 63 | ||
| BC058941.1 | - | 11 - 12 | 1535 | 6825758 | 63 | |
More Information
Set Membership: |
|

Patl1
Genomic Map