Assay Details
| Gene Symbol: Patl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK154102.1 | - | 12 - 13 | 1681 | 145 | |
| AK170918.1 | - | 12 - 13 | 1684 | 145 | ||
| BC047458.1 | - | 12 - 13 | 1485 | 145 | ||
More Information
Set Membership: |
|

Patl1
Genomic Map