Assay Details
| Gene Symbol: Patl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172635.3 | NP_766223.2 | 4 - 5 | 589 | 66 | |
| GenBank mRNA | AK085013.1 | - | 4 - 5 | 531 | 66 | |
| AK087559.1 | - | 4 - 5 | 553 | 66 | ||
| AK150301.1 | - | 4 - 5 | 590 | 66 | ||
| AK154102.1 | - | 4 - 5 | 587 | 66 | ||
| AK170408.1 | - | 4 - 5 | 553 | 66 | ||
| AK170745.1 | - | 4 - 5 | 554 | 66 | ||
| AK170918.1 | - | 4 - 5 | 590 | 66 | ||
| BC047458.1 | - | 3 - 4 | 317 | 66 | ||
| BC058941.1 | - | 4 - 5 | 530 | 6825758 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Patl1
Genomic Map