Assay Details
| Gene Symbol: Gif | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008118.3 | NP_032144.2 | 2 - 3 | 341 | 75 | |
| GenBank mRNA | AK078933.1 | - | 2 - 3 | 342 | 75 | |
| BC118519.1 | - | 2 - 3 | 327 | 40105765 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gif
Genomic Map