Assay Details
| Gene Symbol: Ms4a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276398.1 | NP_001263327.1 | 1 - 2 | 382 | 64 | |
| GenBank mRNA | AK079888.1 | - | 1 - 2 | 383 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ms4a7
Genomic Map