Assay Details
| Gene Symbol: Stx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025307.1 | NP_001020478.1 | 4 - 5 | 971 | 68 | |
| NM_001286543.1 | NP_001273472.1 | 5 - 6 | 1017 | 68 | ||
| NM_011502.3 | NP_035632.1 | 4 - 5 | 917 | 68 | ||
| NM_152220.2 | NP_689344.1 | 4 - 5 | 971 | 68 | ||
| GenBank mRNA | AK038415.1 | - | 4 - 5 | 478 | 68 | |
| AK133830.1 | - | 4 - 5 | 494 | 68 | ||
| AK170105.1 | - | 4 - 5 | 380 | 68 | ||
| AK171135.1 | - | 4 - 5 | 514 | 68 | ||
| BC024844.1 | - | 4 - 5 | 379 | 5357204 | 68 | |
| BC056949.1 | - | 5 - 6 | 1017 | 68 | ||
| D29797.1 | - | 4 - 5 | 392 | 68 | ||
| D29798.1 | - | 4 - 5 | 390 | 68 | ||
| D29799.1 | - | 4 - 5 | 405 | 68 | ||
| D29800.1 | - | 5 - 6 | 349 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001286543.1 transcripts.
Set Membership: |
|

Stx3
Genomic Map