Assay Details
| Gene Symbol: Stx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025307.1 | NP_001020478.1 | 5 - 6 | 1041 | 88 | |
| NM_001286543.1 | NP_001273472.1 | 6 - 7 | 1087 | 88 | ||
| NM_011502.3 | NP_035632.1 | 5 - 6 | 987 | 88 | ||
| NM_152220.2 | NP_689344.1 | 5 - 6 | 1041 | 88 | ||
| GenBank mRNA | AK038415.1 | - | 5 - 6 | 548 | 88 | |
| AK133830.1 | - | 5 - 6 | 564 | 88 | ||
| AK170105.1 | - | 5 - 6 | 450 | 88 | ||
| AK171135.1 | - | 5 - 6 | 584 | 88 | ||
| BC024844.1 | - | 5 - 6 | 449 | 5357204 | 88 | |
| BC056949.1 | - | 6 - 7 | 1087 | 88 | ||
| D29797.1 | - | 5 - 6 | 462 | 88 | ||
| D29798.1 | - | 5 - 6 | 460 | 88 | ||
| D29799.1 | - | 5 - 6 | 475 | 88 | ||
| D29800.1 | - | 6 - 7 | 419 | 88 | ||
More Information
Set Membership: |
|

Stx3
Genomic Map