Assay Details
Gene Symbol: Ms4a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023529.2 | NP_076018.2 | 2 - 3 | 244 | 62 | |
GenBank mRNA | AB026046.1 | - | 2 - 3 | 253 | 62 | |
AF237918.1 | - | 2 - 3 | 244 | 62 | ||
BC144935.1 | - | 2 - 3 | 241 | 9053483 | 62 | |
BC150675.1 | - | 2 - 3 | 241 | 9087586 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
