Assay Details
| Gene Symbol: Mmp17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011846.4 | NP_035976.3 | 8 - 9 | 1295 | 76 | |
| GenBank mRNA | AB021224.2 | - | 8 - 9 | 1301 | 76 | |
| AJ010731.1 | - | 8 - 9 | 1299 | 76 | ||
| AK079221.1 | - | 1067 | 76 | |||
| AK143521.1 | - | 8 - 9 | 1295 | 76 | ||
| BC051917.1 | - | 8 - 9 | 1295 | 5716458 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mmp17
Genomic Map