Assay Details
| Gene Symbol: Cct6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009838.2 | NP_033968.1 | 1423 | 78 | ||
| GenBank mRNA | AK077916.1 | - | 1377 | 78 | ||
| AK134428.1 | - | 1376 | 78 | |||
| AK145522.1 | - | 1383 | 78 | |||
| AK147130.1 | - | 1420 | 78 | |||
| AK149757.1 | - | 1782 | 78 | |||
| AK150977.1 | - | 1377 | 78 | |||
| AK159354.1 | - | 1377 | 79 | |||
| AK159785.1 | - | 1397 | 78 | |||
| AK159975.1 | - | 1423 | 78 | |||
| AK160067.1 | - | 1377 | 79 | |||
| AK164312.1 | - | 1377 | 78 | |||
| AK165612.1 | - | 1379 | 78 | |||
| AK166749.1 | - | 1377 | 78 | |||
| AK166795.1 | - | 1407 | 78 | |||
| AK166870.1 | - | 1375 | 78 | |||
| AK166887.1 | - | 1376 | 78 | |||
| AK166929.1 | - | 1375 | 78 | |||
| AK167007.1 | - | 1405 | 78 | |||
| AK167076.1 | - | 1374 | 78 | |||
| AK167082.1 | - | 1373 | 78 | |||
| AK167164.1 | - | 1382 | 78 | |||
| AK167180.1 | - | 1409 | 78 | |||
| AK167307.1 | - | 1377 | 78 | |||
| AK167669.1 | - | 1406 | 78 | |||
| AK167851.1 | - | 1378 | 78 | |||
| AK167994.1 | - | 1372 | 78 | |||
| AK168065.1 | - | 1372 | 78 | |||
| AK168940.1 | - | 1406 | 78 | |||
| AK169033.1 | - | 1356 | 78 | |||
| BC018459.1 | - | 987 | 3597455 | 78 | ||
| BC055043.1 | - | 1385 | 78 | |||
| BC069917.1 | - | 1378 | 6394714 | 78 | ||
| BC094352.1 | - | 1349 | 6809215 | 78 | ||
| Z31557.1 | - | 1394 | 78 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Cct6a
Genomic Map