Assay Details
| Gene Symbol: Cct6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009838.2 | NP_033968.1 | 1606 | 76 | ||
| GenBank mRNA | AK077916.1 | - | 1560 | 76 | ||
| AK134428.1 | - | 1559 | 76 | |||
| AK145522.1 | - | 1566 | 76 | |||
| AK147130.1 | - | 1603 | 76 | |||
| AK149757.1 | - | 1965 | 76 | |||
| AK150977.1 | - | 1560 | 76 | |||
| AK159354.1 | - | 1561 | 76 | |||
| AK159785.1 | - | 1580 | 76 | |||
| AK159975.1 | - | 1606 | 76 | |||
| AK160067.1 | - | 1561 | 76 | |||
| AK164312.1 | - | 1560 | 76 | |||
| AK165612.1 | - | 1562 | 76 | |||
| AK166749.1 | - | 1560 | 76 | |||
| AK166795.1 | - | 1590 | 76 | |||
| AK166870.1 | - | 1558 | 76 | |||
| AK166887.1 | - | 1559 | 76 | |||
| AK166929.1 | - | 1558 | 76 | |||
| AK167007.1 | - | 1588 | 76 | |||
| AK167076.1 | - | 1557 | 76 | |||
| AK167082.1 | - | 1556 | 76 | |||
| AK167164.1 | - | 1565 | 76 | |||
| AK167180.1 | - | 1592 | 76 | |||
| AK167307.1 | - | 1560 | 76 | |||
| AK167669.1 | - | 1589 | 76 | |||
| AK167851.1 | - | 1561 | 76 | |||
| AK167994.1 | - | 1555 | 76 | |||
| AK168065.1 | - | 1555 | 76 | |||
| AK168940.1 | - | 1589 | 76 | |||
| AK169033.1 | - | 1539 | 76 | |||
| BC018459.1 | - | 1170 | 3597455 | 76 | ||
| BC055043.1 | - | 1568 | 76 | |||
| BC069917.1 | - | 1561 | 6394714 | 76 | ||
| BC094352.1 | - | 1532 | 6809215 | 76 | ||
| Z31557.1 | - | 1577 | 76 | |||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24090829 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24090827 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24090828 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24090830 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2125912 | BC094352.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2125912 | Z31557.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2125912 | BC018459.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2125912 | BC055043.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2125912 | BC069917.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 601377 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 971272 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 501549 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 250492 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 969819 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 5175 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 93538 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P116736 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2125912 | BC094352.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2125912 | Z31557.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2125912 | BC018459.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2125912 | BC055043.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2125912 | BC069917.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24090866 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24090864 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24090840 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24090851 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24090829 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24090848 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24090882 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24090884 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24090844 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24090853 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24090846 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24090888 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24090836 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24090837 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24090827 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24090885 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24090838 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24090860 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24090831 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24090886 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24090856 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24090890 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24090865 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24090832 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24090857 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24090893 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24090861 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24090834 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24090842 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24090868 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24090835 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24090828 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24090855 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24090881 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P116736 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05743099 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05743100 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01220245 | - | - |
| Whole Mouse Genome G4122A | A_52_P363718 | BC069917.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P363718 | BC094352.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P363718 | Z31557.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P363718 | BC018459.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P363718 | BC055043.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P104251 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P116736 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05743099 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05743100 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04394009 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04864531 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04864539 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04958578 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04864530 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04394011 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04394013 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04864537 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04958575 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04864536 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04394012 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04864529 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24090828 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24090829 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24090830 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24090827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24090836 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090864 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090844 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090853 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04958580 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090827 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090875 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090840 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090834 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090861 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090842 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090858 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090837 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090886 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090873 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090871 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090893 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090856 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090855 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090884 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090846 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090888 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090849 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090868 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090836 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090890 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04115863 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090829 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04958575 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090865 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090881 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090832 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090851 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090835 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090879 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090880 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090860 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090876 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090831 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04831306 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04864530 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090885 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24090838 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10526084 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526082 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526071 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526080 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526081 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526073 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526075 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526072 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526083 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526077 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526079 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526078 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526076 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526074 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10526070 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423517_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455988_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10526069 | - | - |
| Mouse Genome 430 2.0 Array | 1423517_at | - | - |
| Mouse Genome 430 2.0 Array | 1455988_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1423517_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1455988_a_at | - | - |
| Mouse Expression Set 430 | 1423517_at | - | - |
| Mouse Expression Set 430 | 1455988_PM_a_at | - | - |
| Mouse Expression Set 430 | 1423517_PM_at | - | - |
| Mouse Expression Set 430 | 1455988_a_at | - | - |
| Murine Genome U74v2 Set | 160459_PM_r_at | - | - |
| Murine Genome U74v2 Set | 160459_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423517_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423517_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455988_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455988_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423517_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455988_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455988_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423517_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2350368 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002350368 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2774942 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2774942 | - | - |
| mouse-6 v1.1 (Illumina) | scl27178.12.102_2-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2350368 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl27178.12.102_2-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2350368 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2962840 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2774941 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2962840 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl27178.12.102_2-S | - | - |

Cct6a
Genomic Map