Assay Details
| Gene Symbol: Cct6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009838.2 | NP_033968.1 | 420 | 120 | ||
| GenBank mRNA | AK077916.1 | - | 373 | 120 | ||
| AK134428.1 | - | 373 | 120 | |||
| AK145522.1 | - | 379 | 120 | |||
| AK147130.1 | - | 417 | 120 | |||
| AK149757.1 | - | 779 | 120 | |||
| AK150977.1 | - | 374 | 120 | |||
| AK159354.1 | - | 374 | 120 | |||
| AK159785.1 | - | 394 | 120 | |||
| AK159975.1 | - | 420 | 120 | |||
| AK160067.1 | - | 374 | 120 | |||
| AK164312.1 | - | 373 | 120 | |||
| AK165612.1 | - | 376 | 120 | |||
| AK166749.1 | - | 373 | 120 | |||
| AK166795.1 | - | 403 | 120 | |||
| AK166870.1 | - | 371 | 120 | |||
| AK166887.1 | - | 372 | 120 | |||
| AK166929.1 | - | 372 | 120 | |||
| AK167007.1 | - | 402 | 120 | |||
| AK167076.1 | - | 371 | 120 | |||
| AK167082.1 | - | 369 | 120 | |||
| AK167164.1 | - | 379 | 120 | |||
| AK167180.1 | - | 406 | 120 | |||
| AK167307.1 | - | 374 | 120 | |||
| AK167669.1 | - | 403 | 120 | |||
| AK167851.1 | - | 374 | 120 | |||
| AK167994.1 | - | 369 | 120 | |||
| AK168065.1 | - | 369 | 120 | |||
| AK168940.1 | - | 403 | 120 | |||
| AK169033.1 | - | 353 | 120 | |||
| AK216762.1 | - | 261 | 120 | |||
| BC055043.1 | - | 382 | 120 | |||
| BC069917.1 | - | 375 | 6394714 | 120 | ||
| BC094352.1 | - | 346 | 6809215 | 120 | ||
| Z31557.1 | - | 391 | 120 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Cct6a
Genomic Map