Assay Details
| Gene Symbol: Cct6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009838.2 | NP_033968.1 | 600 | 80 | ||
| GenBank mRNA | AK077916.1 | - | 553 | 80 | ||
| AK134428.1 | - | 553 | 80 | |||
| AK145522.1 | - | 559 | 80 | |||
| AK147130.1 | - | 597 | 80 | |||
| AK149757.1 | - | 959 | 80 | |||
| AK150977.1 | - | 554 | 80 | |||
| AK159354.1 | - | 554 | 80 | |||
| AK159785.1 | - | 574 | 80 | |||
| AK159975.1 | - | 600 | 80 | |||
| AK160067.1 | - | 554 | 80 | |||
| AK164312.1 | - | 553 | 80 | |||
| AK165612.1 | - | 556 | 80 | |||
| AK166749.1 | - | 553 | 80 | |||
| AK166795.1 | - | 583 | 80 | |||
| AK166870.1 | - | 551 | 80 | |||
| AK166887.1 | - | 552 | 80 | |||
| AK166929.1 | - | 552 | 80 | |||
| AK167007.1 | - | 582 | 80 | |||
| AK167076.1 | - | 551 | 80 | |||
| AK167082.1 | - | 549 | 80 | |||
| AK167164.1 | - | 559 | 80 | |||
| AK167180.1 | - | 586 | 80 | |||
| AK167307.1 | - | 554 | 80 | |||
| AK167669.1 | - | 583 | 80 | |||
| AK167851.1 | - | 554 | 80 | |||
| AK167994.1 | - | 549 | 80 | |||
| AK168065.1 | - | 549 | 80 | |||
| AK168940.1 | - | 583 | 80 | |||
| AK169033.1 | - | 533 | 80 | |||
| AK204878.1 | - | 49 | 80 | |||
| BC018459.1 | - | 164 | 3597455 | 80 | ||
| BC055043.1 | - | 562 | 80 | |||
| BC069917.1 | - | 555 | 6394714 | 80 | ||
| BC094352.1 | - | 526 | 6809215 | 80 | ||
| Z31557.1 | - | 571 | 80 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Cct6a
Genomic Map